Order Entry
Belgium
Orders LinkContactUsLinkComponent
TASKI® Sani Calc W3b Sanitary Basic Cleaner

TASKI® Sani Calc W3b Sanitary Basic Cleaner

W3b 2×5 L Sanitary basic cleaner - descaler, 198W×287L×309H mm, 5 L, 2 pH, Box
Catalog # DIVY101108863
Supplier:   Diversey
TASKI® Sani Calc W3b Sanitary Basic Cleaner
Catalog # DIVY101108863
Supplier:  Diversey
TASKI® Sani Calc W3b Sanitary Basic Cleaner

W3b 2×5 L Sanitary basic cleaner - descaler, 198W×287L×309H mm, 5 L, 2 pH, Box

Price
55,50 €
Log in for availability
1 * 2 Fl
Product Details & Documents

Powerful descaler for daily and periodic cleaning and descaling of acid-resistant sanitary surfaces.

  • Based on phosphoric acid
  • Integrated corrosion inhibitors
  • Fast acting
  • Unique, patented odor neutralization technology (ONT)
  • Strong lime dissolving power for quick removal of stubborn lime deposits
  • Gentle cleaning
  • Excellent for efficient, comprehensive limescale removal from floors, walls and other sanitary surfaces
  • Leaves a pleasant, fresh scent

TASKI® Sani Calc is dosed either manually or with a pump. Correct dosing saves costs and minimizes environmental impact.

Specifications
  • Capacity:
    5 L
  • Description:
    W3b 2×5 L Sanitary basic cleaner - descaler, 198W×287L×309H mm
  • Pack type:
    Box
  • pH value:
    2 pH
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.