Order Entry
Belgium
ContactUsLinkComponent
Pepsin Porcine
Pepsin Porcine
Catalog # ENZOSV-31820-01
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Pepsin Porcine
Catalog # ENZOSV-31820-01
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    1 g
  • CAS No.:
    9001-75-6
  • Storage conditions:
    −20 °C
  • Shipping conditions:
    Ambient temperature
  • Enzyme name:
    Pepsin porcine
  • Enzyme activity:
    15 milli Anson units/mg

Specifications

About this item

Caution: Research use only.