CleanMax® Clean Manufactured Coveralls with Attached Hood and Boots
Supplier: LAKELAND CLEANMAX APPAREL
New Product
Certificates
About this item
Cleanroom garments are constructed and packaged to offer premium protection for your cleanroom and controlled environments.
- Attached hood
- Attached boots
- Elastic wrist
Cleanroom coveralls with attached hood and boots is ideal for ISO Class 4 to 6 cleanrooms.
Packaging: These products are offered in a case of 25.
Specifications
- Catalog No:
- 114-9576
- CLLLCTL414CM-SM
- 114-9574
- 114-9575
- 114-9572
- 114-9573
- 114-9570
- 114-9571
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Seam type:
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Bound
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom