Orders LinkContactUsLinkComponent
UTTD Control Pharma Solution Homogenising System
UTTD Control Pharma Solution Homogenising System
UTTD control pharma solution
Catalog # IKAA10013297
Supplier:   IKA
UTTD Control Pharma Solution Homogenising System
Catalog # IKAA10013297
Supplier:  IKA
UTTD Control Pharma Solution Homogenising System
UTTD Control Pharma Solution Homogenising System
UTTD control pharma solution
Price
1 730,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Many standardised test procedures in the pharmaceutical industry, such as the pharmacopoeia tests described in official compendia (United States Pharmacopeia (USP 905), European Pharmacopoeia (PhEur 2.9.40), etc.), require the crushing and dissolution of tablets as a first step before further analysis. The challenges in sample preparation include the avoidance of cross-contamination between sample batches, time-consuming cleaning and the often small sample quantities.
  • Crushing and dissolving of tablets for subsequent analysis
The package includes the powerful ULTRA-TURRAX® Tube Drive P control homogenising system, which has been designed for universal use. In combination with the disposable mixing vessels with dissolver disc (DIS-300-S-M with 300 ml volume), this system ensures optimum comminution and thorough dissolution of tablets and capsules in various solvents.
The hermetically sealable disposable sample containers serve simultaneously as a comminution vessel, storage unit and transport container. They significantly reduce the cleaning effort and systematically eliminate cross-contamination. Users benefit from reduced handling costs and a lower number of incorrect analyses. Samples can be collected or media added via the pierceable membrane in the vessel lid.
Delivery information: ULTRA-TURRAX® Tube Drive P control, DIS-300-S-M.10 Dissolver tube, 300 ml, 10 pieces.
Specifications
  • Description:
    UTTD control pharma solution
  • Cat. No.:
    IKAA10013297
  • Speed display:
    OLED
  • Speed range:
    400 - 8000 min⁻¹
  • Dimensions:
    122×178×54 mm
  • Max. viscosity:
    5000 mPas
  • Power rating:
    36 W
  • Weight:
    1,3 kg
  • Motor rating input/output:
    36/28 W
Accessories
Product Information
  • Catalog #
    Volume
    Closure type
    Availability
    Price
  • ST-50, tube with stirring device
    ST-50, tube with stirring device
    531-0480
    50 ml
    N
    Log in for availability
    1 * 10 ST - 114,00 €
Product Information
  • Catalog #
    Volume
    Closure type
    Availability
    Price
  • BMT-20-S, tube for milling with stainless steel balls
    BMT-20-S, tube for milling with stainless steel balls
    531-0469
    20 ml
    N
    Log in for availability
    1 * 25 ST - 251,00 €
Product Information
  • Catalog #
    Volume
    Closure type
    Availability
    Price
  • BMT-20-G tube for milling with glass balls
    BMT-20-G tube for milling with glass balls
    431-2893
    20 ml
    N
    Log in for availability
    1 * 25 ST - 251,00 €
Product Information
  • Catalog #
    Volume
    Closure type
    Availability
    Price
  • DT-50, tube with rotor/stator element
    DT-50, tube with rotor/stator element
    531-0481
    50 ml
    N
    Log in for availability
    1 * 10 ST - 128,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Glass beads for BMT tubes, Ø 6 mm
    Glass beads for BMT tubes, Ø 6 mm
    IKAA3599200
    Log in for availability
    1 * 250 g - 69,00 €
Product Information
  • Catalog #
    Volume
    Closure type
    Availability
    Price
  • BMT-50-S-M-gamma, tube for milling with stainless steel balls
    BMT-50-S-M-gamma, tube for milling with stainless steel balls
    531-0493
    50 ml
    M
    Log in for availability
    1 * 10 ST - 218,00 €
Product Information
  • Catalog #
    Volume
    Closure type
    Availability
    Price
  • BMT-50-G, tube for milling with glass balls
    BMT-50-G, tube for milling with glass balls
    531-0483
    50 ml
    N
    Log in for availability
    1 * 10 ST - 154,00 €
Product Information
  • Catalog #
    Volume
    Closure type
    Availability
    Price
  • ST-20-M-gamma, tube with stirring device
    ST-20-M-gamma, tube with stirring device
    531-0477
    20 ml
    M
    Log in for availability
    1 * 20 ST - 255,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Labworldsoft® 6 Starter
    Labworldsoft® 6 Starter
    IKAA20019397
    Small configurations with up to 3 devices can be controlled at the same time
    Log in for availability
    1 * 1 ST - 6 500,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Labworldsoft® 6 Pro, with access to the entire device library
    Labworldsoft® 6 Pro, with access to the entire device library
    IKAA20017366
    All configurations with any number of devices can be controlled at the same time
    Log in for availability
    1 * 1 ST - 14 820,00 €
Product Information
  • Catalog #
    Volume
    Closure type
    Availability
    Price
  • BMT-20-S-M-gamma, tube for milling with stainless steel balls
    BMT-20-S-M-gamma, tube for milling with stainless steel balls
    IKAA3700701
    20 ml
    M
    Log in for availability
    1 * 20 ST - 293,00 €
Product Information
  • Catalog #
    Volume
    Closure type
    Availability
    Price
  • BMT-50-S, tube for milling with stainless steel balls
    BMT-50-S, tube for milling with stainless steel balls
    531-0482
    50 ml
    N
    Log in for availability
    1 * 10 ST - 154,00 €
Product Information
  • Catalog #
    Volume
    Closure type
    Availability
    Price
  • DT-20 eco, tube with rotor/stator element
    DT-20 eco, tube with rotor/stator element
    IKAA20003207
    N
    Log in for availability
    1 * 25 ST - 204,00 €
Product Information
  • Catalog #
    Volume
    Closure type
    Availability
    Price
  • ST-50-M-gamma, tube with stirring device
    ST-50-M-gamma, tube with stirring device
    531-0491
    50 ml
    M
    Log in for availability
    1 * 10 ST - 180,00 €
Product Information
  • Catalog #
    Availability
    Price
  • TC-20-M, closure for 20 ml tubes with pierceable membrane
    TC-20-M, closure for 20 ml tubes with pierceable membrane
    531-0496
    Log in for availability
    1 * 25 ST - 70,00 €
Product Information
  • Catalog #
    Volume
    Closure type
    Availability
    Price
  • DT-50-M-gamma, tube with rotor/stator element
    DT-50-M-gamma, tube with rotor/stator element
    531-0492
    50 ml
    M
    Log in for availability
    1 * 10 ST - 204,00 €
Product Information
  • Catalog #
    Volume
    Closure type
    Availability
    Price
  • DT-20-M-gamma, tube with rotor/stator element
    DT-20-M-gamma, tube with rotor/stator element
    531-0478
    20 ml
    M
    Log in for availability
    1 * 20 ST - 281,00 €
Product Information
  • Catalog #
    Volume
    Closure type
    Availability
    Price
  • ST-20, tube with stirring device
    ST-20, tube with stirring device
    431-2892
    20 ml
    N
    Log in for availability
    1 * 25 ST - 228,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Stainless steel (AISI 304) for BMT tubes, Ø 5 mm
    Stainless steel (AISI 304) for BMT tubes, Ø 5 mm
    IKAA3599300
    Log in for availability
    1 * 250 g - 69,00 €
Product Information
  • Catalog #
    Availability
    Price
  • TC-50-M, closure for 50 ml tubes with pierceable membrane
    TC-50-M, closure for 50 ml tubes with pierceable membrane
    431-0165
    Log in for availability
    1 * 10 ST - 46,00 €
Product Information
  • Catalog #
    Volume
    Closure type
    Availability
    Price
  • DT-20, tube with rotor/stator element
    DT-20, tube with rotor/stator element
    431-0135
    20 ml
    N
    Log in for availability
    1 * 25 ST - 237,00 €