ContactUsLinkComponent
VWR® Compact Vacuum Pumps For Filtration and Solid Phase Extraction
VWR® Compact Vacuum Pumps For Filtration and Solid Phase Extraction
Supplier:  VACUUBRAND
Certificates
undefined
VWR® Compact Vacuum Pumps For Filtration and Solid Phase Extraction
Supplier:  VACUUBRAND
Specifications
  • Catalog No:
  • 181-0238
  • 181-0235
  • Model:
  • VP 100 C
  • VP 100
  • No. of stages:
  • 1
  • 1
  • Flow rate:
  • 0,7 m³/h
  • 0,7 m³/h
  • Vacuum:
  • 100 mbar
  • 100 mbar