Order Entry
Belgium
ContactUsLinkComponent
Ethylenetetracarbonitrile
Toxic
Ethylenetetracarbonitrile
Catalog # MAYBBTB10306.1
Supplier:  Thermo Scientific
CAS Number:  
Toxic
Ethylenetetracarbonitrile
Catalog # MAYBBTB10306.1
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :128.09 g/mol
Merck Index:12,09336
Boiling Pt:223 °C (1013 hPa)
Melting Pt:196…199 °C
Density:1.35 g/cm³ (25 °C)
EINECS:211-578-0
UN:3439
ADR:6.1,II
 :GHS06
CAS Number:670-54-2
Formula:C₆N₄
MDL Number:MFCD00001850