Order Entry
Belgium
ContactUsLinkComponent
 
Corrosive
Sulphuric acid 1 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1086804
Catalog # 85973.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Sulphuric acid 1 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1086804
Catalog # 85973.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:3264
ADR:8,III
 :GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589