Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid 0.9 mol/l (1.8 N) in aqueous solution
Irritant
Sulphuric acid 0.9 mol/l (1.8 N) in aqueous solution
Catalog # KRAF06187.7900
Supplier:  Bernd Kraft
CAS Number:  
Irritant
Sulphuric acid 0.9 mol/l (1.8 N) in aqueous solution
Catalog # KRAF06187.7900
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    25 L
  • Seq:
    5327

Specifications

About this item

 :
 :98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
 :GHS07
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589