Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid concentrate for dilution to 0.01 mol, ConvoL NORMADOSE®
Sulphuric acid concentrate for dilution to 0.01 mol, ConvoL NORMADOSE®
Catalog # 83760.600
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sulphuric acid concentrate for dilution to 0.01 mol, ConvoL NORMADOSE®
Catalog # 83760.600
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    60 mL
  • Seq:
    2385

Specifications

About this item

 :
 :98.08 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2796
ADR:8,II
 :Dose for preparation of 1 litre of solution.
EINECS:231-639-5
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589