Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid 0.005 mol/l concentrated aqueous solution, Supelco®
Sulphuric acid 0.005 mol/l concentrated aqueous solution, Supelco®
Catalog # 1.09982.0001
Supplier:  Merck
CAS Number:  
Sulphuric acid 0.005 mol/l concentrated aqueous solution, Supelco®
Catalog # 1.09982.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.01 g/cm3 (20 °C)
  • pH:
    1.1 (H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
 :98.08 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3264
ADR:8,
 :M-Clarity™ quality level = MQ100
EINECS:231-639-5
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589