Methyl-3-nitrosalicylate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:22621-41-6
:
Formula:C₈H₇NO₅
MDL Number:MFCD00272263
:197.15 g/mol
Melting Pt:129…132 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together