Neocuproine hydrochloride monohydrate, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: 2,9-Dimethyl-1,10-phenanthroline hydrochloride
CAS Number:303136-82-5
:
Formula:C₁₄H₁₃ClN₂·1H₂O
MDL Number:MFCD00012645
Molecular Weight:262.74 g/mol
:M-Clarity™ quality level = MQ100
EINECS:230-732-8
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together