Order Entry
Belgium
ContactUsLinkComponent
Toluene-[D₈] (99.5% D) for NMR spectroscopy
Toluene-[D₈] (99.5% D) for NMR spectroscopy
Catalog # 87159.0010
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toluene-[D₈] (99.5% D) for NMR spectroscopy
Catalog # 87159.0010
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.9 %
  • Isotopic enrichment (FT NMR 400 MHz)(D):
    Min. 99.5 %
  • Water (HDO+D2O):
    Max. 0.0200 %

Specification Test Results

About this item

 :
 :100.08 g/mol
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–85 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:4 °C (closed cup)
Storage Temperature:Refrigerator
EINECS:218-009-5
UN:1294
ADR:3,II
 :GHS02
 :GHS08
 :GHS06
CAS Number:2037-26-5
Formula:C₆D₅CD₃
MDL Number:MFCD00044638