Paraplast® for tissue, for embedding
Catalog # P3558-1KG
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Note:Paraplast is a registered trademark of Leica Biosystems
- Moisture content
< or = 0.2%:Conforms
Specification Test Results
About this item
:
Melting Pt:56...57 °C (lit.)
:M-Clarity™ quality level = MQ200
Storage Temperature:Room Temperature
CAS Number:8002-74-2
:8042-47-5