TISSUE-TROL™ Control Slides Argentaffin
Catalog # TTR013-25EA
Supplier: SIGMA ALDRICH MICROSCOPY
Specifications
- Description:Argentaffin TISSUE-TROL™ control slides from human intestine tissue containing argentaffin granules
- Pk:25
Specifications
About this item
Histology slides with paraffin embedded section of human intestine containing argentaffin granules. Suitable for use with Fontana-Masson stain kit (Procedure No. HT200).
Delivery information: Box contains 24 unstained slides plus one reference slide.