Order Entry
Belgium
Orders LinkContactUsLinkComponent
Wright-Giemsa stain modified

Wright-Giemsa stain modified

4 L
Catalog # WG128-4L
Inflammable
Toxique
Santé
Wright-Giemsa stain modified
Catalog # WG128-4L
CAS Number:  
Wright-Giemsa stain modified

4 L

CAS Number:  
Price
563,00 €
Connexion Pour la disponibilité
1 * 4 L
Product Details & Documents

When blood films are stained using Wright-Giemsa Stain, the white blood cell nucleus and cytoplasm take on the characteristic blue or pink coloration. The combination of purified eosin and thiazine dyes in the product eliminates inconsistent staining and yields reproducible chromogenic responses.

Synonymes:  Wright-Giemsa Solution
  • Numéro MDL:MFCD00132593
  •  :M-Clarity™ quality level = MQ500 (more info.)
  • Point d'éclair:11 °C (closed cup)
  • Température de stockage:Room Temperature

Documents

Spécifications

Résultats des tests de spécification

  • Appearance (Turbidity)
    Essentially free of precipitate:
    Clear
  • Appearance (Colour):
    Blue
  • Appearance (Form):
    Solution
  • pH:
    6.7 - 6.9
  • 18 months:
    Conforms
    Cells stain within the performance characteristics when using Sigma-Aldrich Procedure No. WG

Spécifications

  • Pk:
    4 L

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.