
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride ≥95%, powder
Catalog # T3134-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance (Colour):Off-White to Grey to Brown
- Appearance (Form):Powder
- Infrared Spectrum:Conforms to Structure
- Purity (HPLC):≥95 %
- Titration with HClO₄:≥99 %
- Solubility (Coloru):Colorless to Light Purple to Dark Brown
- Solubility (Turbidity)
5 g/100 mL, H2O:Clear to Very Slightly Hazy - Recommended Retest Period:2 Years
Specification Test Results
About this item
:
:237.17 g/mol
Melting Pt:222...224 °C (lit.)
:M-Clarity™ quality level = MQ200
:GHS07
Storage Temperature:Room Temperature
CAS Number:637-01-4
Formula:C₁₀H₁₈Cl₂N₂
MDL Number:MFCD00012482