Orders LinkContactUsLinkComponent
Avantor is setting science in motion for a better world | Avantor
Poly prep slides
Catalog # P0425-72EA
Catalog # P0425-72EA
Poly prep slides
Price
120,00 €
Log in for availability
1 * 72 ST
Product Details & Documents
Poly-prep slides are made of glass and designed for cell adhesion. The slides are coated with Poly-L-Lysine, a non-specific attachment factor for cells useful in promoting cell adhesion on solid substrates. Poly-L-lysine enhances binding between the cell membrane and the cultured surface.
  • Poly-Prep Slides are used:
  • To mount liver and lung cell sections and mounted on poly-prep slides
  • To prove better cell adhesion in clinical bronchoalveolar lavage fluid samples
  • To determine pAkt (alpha serine/threonine-protein kinase) expression by immunohistochemistry
  • In immunofluorescence imaging of nuclear translocation of nuclear factor κB(NFκB) in A549 cells
  • To study the phospholipase C-zeta activity of sperm
  • To prepare coelomocytes smears, which were further subjected to antibody staining
  • Hematology, histology
Overall, glass slides coated with poly-L-lysine for tissue section adhesion in routine histological uses. And, it is not recommended for cell culture or microarray applications.
Specifications
  • Description:
    Poly prep slides