Order Entry
Belgium
Orders LinkContactUsLinkComponent
Phosphate Buffer, pH 6.6 at 25 °C

Phosphate Buffer, pH 6.6 at 25 °C

Phosphate Buffer, pH 6.6 at 25 °C
Certificates

2 Options Available Below

Product Details & Documents

Phosphate buffer's buffering capacity is pH sensitive and is preferred to be used at physiological pH (7,2 to 7,4).

  • Each vial will prepare 3,8 L of buffer, 34 mM
  • Used in hematology and histology

For use with Wright Stain for Geometric Data HEMASTAIN®. Phosphate buffer serves as an important fixative for light and electron microscopy. It is the most commonly used component of irrigation solution during surgeries, to preserve living tissues. It is also used in immunochemical staining procedures.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • P8165-1VL
  • P8165-12VL
  • Appearance:
  • White powder
  • White powder
  • Storage temperature:
  • RT
  • RT
  • M-Clarity quality level:
  • MQ200
  • MQ200

Specifications

Product Family Options

Product Information

  • pH value
    Pack type
    Pk
    Availability
    Price
  • P8165-1VL
    Phosphate buffer
    Phosphate buffer
    6,6
    1 vial in poly bottle
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 115,00 €
    115,00 €
    1 * 1 ST
       115,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    P8165-1VL
    Appearance
    White powder
    Storage temperature
    RT
    M-Clarity quality level
    MQ200
  • P8165-12VL
    Phosphate buffer
    Phosphate buffer
    6,6
    12
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 283,00 €
    283,00 €
    1 * 12 ST
       283,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    P8165-12VL
    Appearance
    White powder
    Storage temperature
    RT
    M-Clarity quality level
    MQ200
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.