Order Entry
Belgium
ContactUsLinkComponent
 
Nunc™ Bioreactor Centrifuge Tube
Catalog # 525-1611
Nunc™ Bioreactor Centrifuge Tube
Catalog # 525-1611

Specifications

  • Sterile:
    +
  • Capacity:
    50 ml
  • Description:
    Bioreactor centrifuge tube
  • Material:
    PP
  • Cap type:
    HDPE, quadruple-position vent/close cap
  • Pk:
    90
  • Tube bottom shape:
    Conical

Specifications

About this item

Thermo Scientific™ Nunc™ Bioreactor centrifuge tubes are ideal for screening, small volume high throughput analysis.

  • Protein expression suspension culture volumes ranging from 10 to 25 ml
  • Convenient pack sizes
  • Four different sizes of ventilation (dial type) on the cap
  • Tube: polypropylene, cap: HDPE
  • Filter: PTFE, dial: polystyrene
  • 2 μm PTFE filter membrane
  • Suitable for suspension culture
  • Sterile - SAL 10⁻⁶, non-pyrogenic and non-cytotoxic
  • Shelf life: Two years

Thermo Scientific™ Nunc™ Bioreactor centrifuge tubes are designed for high throughput protein expression screening and cell line development and optimisation under suspension condition. Adjustable 4-level vented cap with a 0,2 μm filter membrane allows for controlled gas exchange and air circulation without contamination. Ergonomic grooved cap for effortless handling and a large writing area for ease of identification.

Ordering information: Supplied DNase/RNase free with HDPE cap, PTFE filter, polystyrene dial and 0,2 μm PTFE filter membrane.
Packaging: 3 per pack, 90 per case.