Orders LinkContactUsLinkComponent
Point Sampler
Point Sampler
Supplier:   Sampling Systems
Point Sampler
Supplier:  Sampling Systems
Certificates
Point Sampler
2 Options Available Below
Product Details & Documents
The point sampler is designed for large samples of powders.
  • Ideal for taking large volumes of powder from a single point
  • Perfect for both free-flowing and sticky powders
  • Easy to operate
  • Available from stock
  • Fast delivery
The point sampler is able to quickly take a single, large volume sample of both free-flowing or cohesive powders. The easy-to-use design means the scraper pushes the product into the sampler when the device is rotated.
Product Family Options
Product Information
  • Material
    Length
    Volume
    Availability
    Price
  • SAMP1640A-1000
    Point sampler
    Point sampler
    Stainless steel 316L
    1000 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 950,00 €
    1 950,00 €
    1 * 1 ST
       1 950,00 €
  • SAMP1640A-2000
    Point sampler
    Point sampler
    Stainless steel 316L
    2000 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 990,00 €
    1 990,00 €
    1 * 1 ST
       1 990,00 €