ContactUsLinkComponent
GenSwab™ for Collection and Preservation of Biological Samples
GenSwab™ for Collection and Preservation of Biological Samples
Catalog # BINZGS2CGSW
Supplier:  Ahlstrom-Munksjö
New Product
GenSwab™ for Collection and Preservation of Biological Samples
Catalog # BINZGS2CGSW
Supplier:  Ahlstrom-Munksjö
New Product
Specifications
  • Sterility:
    Sterile
  • Description:
    GenSwab™ with integrated specimen collection card
  • Material:
    Foam
  • Pk:
    100
Specifications
About this item
GenSwab™ specifically designed for easy collection and long-term preservation of buccal samples to achieve high recovery of DNA and high-quality genetic profiles.
  • Environmentally friendly paper-based design
  • Automated design compatible with automated DBS equipment
  • Complete protection of samples during transport
GenSwab™ allows easy collection and long-term preservation of buccal samples at ambient temperature, safeguarding their quality over time for downstream processing and improving sample traceability with multiple barcodes.
Certifications: The device is manufactured in compliance with the ISO 18385:2016 standard.