Refractive Indexx Liquid, Series AA
Catalog # CARG1806Y-1.4400-8
Supplier: CARGILLE
New Product
CAS Number:
Specifications
- Pk:1
- Refractive index:1,4400 nD
Specifications
About this item
Refractive index liquids have a broad range of applications in diversified fields.
- Colourless formulation
- Stable
- Shelf life unopened: 5 years from date of manufacture
- Shelf life open: Half the remaining time after opening
- Keep tightly capped
Adjustment: ±0,0002