ContactUsLinkComponent
STEMdiff™ Cardiomyocyte Plating Kit
STEMdiff™ Cardiomyocyte Plating Kit
Catalog # STMC100-1121
CAS Number:  
undefined
STEMdiff™ Cardiomyocyte Plating Kit
Catalog # STMC100-1121
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    STEMdiff™ Cardiomyocyte plating kit
Specifications
About this item
A specialised media kit for improving plating efficiency post-thawing and post-harvesting of human pluripotent stem cell-derived cardiomyocytes.
Double the yield of human pluripotent stem cell (hPSC)-derived cardiomyocytes following dissociation, thawing, and replating by using the STEMdiff™ Cardiomyocyte Plating Kit. Consisting of the STEMdiff™ Cardiomyocyte Plating Supplement and STEMdiff™ Cardiomyocyte Support Medium, this kit improves plating efficiency and minimises the loss of hPSC-derived cardiomyocytes. This kit is designed for hPSC-derived cardiomyocytes generated using STEMdiff™ Ventricular Cardiomyocyte Differentiation Kit or STEMdiff™ Atrial Cardiomyocyte Differentiation Kit. By doubling your cardiomyocyte plating efficiency, the STEMdiff™ Cardiomyocyte Plating Kit increases experimental throughput.
Use this kit as part of a complete, optimised workflow including these compatible STEMdiff™ cardiomyocyte products: STEMdiff™ Ventricular Cardiomyocyte Differentiation Kit, STEMdiff™ Atrial Cardiomyocyte Differentiation Kit, STEMdiff™ Cardiomyocyte Dissociation Kit, STEMdiff™ Cardiomyocyte Support Medium, STEMdiff™ Cardiomyocyte Freezing Medium, STEMdiff™ Cardiomyocyte Maintenance Kit.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').