Specifications
- Pk:1 KIT
- Description:STEMdiff™ Microglia differentiation kit
Specifications
About this item
Differentiation kit for the generation of microglia precursors from human ES and iPS cell-derived hematopoietic progenitor cells.
The STEMdiff™ Microglia Culture System comprises STEMdiff™ Microglia Differentiation Kit and STEMdiff™ Microglia Maturation Kit. Together, these kits are used to differentiate and mature microglia derived from human pluripotent stem cells (hPSCs) using STEMdiff™ Hematopoietic Kit. Based on the protocol from the laboratory of Mathew Blurton-Jones, the resulting cells are a highly pure population of microglia (> 80% CD45/CD11b-positive, >50% TREM2-positive microglia; < 20% morphologically distinct monocytes or macrophages). Cells derived using these products are versatile tools for modeling neuroinflammation, studying human neurological development and disease, co-culture applications, and toxicity testing.