MEM Non-Essential Amino Acid Solution (100X)
Catalog # STMC07600
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:100 mL
- Description:MEM Non-Essential amino acid solution (100X)
Specifications
About this item
MEM non-essential amino acid solution (100X).
MEM Non-Essential Amino Acid Solution (100X) has been pre-screened for use with other reagents of the ES-Cult™ product line for optimal performance for the in vitro hematopoietic differentiation of mouse embryonic stem (ES) cells and mouse induced pluripotent stem (iPS) cells.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').