- Pk:2 mL
- Description:ImmunoCult™-ACF Human B cell expansion supplement
Serum-free and animal component-free culture supplement for expansion of human B cells.
ImmunoCult™-ACF Human B Cell Expansion Supplement is a serum-free and animal component-free supplement formulated to promote robust in vitro expansion of human peripheral blood B cells in the absence of serum, feeder cells, or specialized culture plates. The supplement contains recombinant proteins and chemically defined components that support the activation and expansion of human B cells and their maturation to plasma cells. Available either individually or as part of ImmunoCult™ Human B Cell Expansion Kit.
ImmunoCult™-ACF Human B Cell Expansion Supplement is a 50X supplement that can be added to a base medium such as ImmunoCult™-XF B Cell Base Medium to prepare a complete serum-free medium for culturing and expanding human B cells.