ContactUsLinkComponent
STEMdiff™ Erythroid Kit
STEMdiff™ Erythroid Kit
Catalog # STMC100-0074
CAS Number:  
undefined
STEMdiff™ Erythroid Kit
Catalog # STMC100-0074
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    STEMdiff™ Erythroid kit
Specifications
About this item
For differentiation of human ES or iPS cells into erythroid progenitor cells.
STEMdiff™ Erythroid Kit is designed for the serum-free and feeder-free differentiation of human embryonic stem (ES) and induced pluripotent stem (iPS) cells to erythroid progenitor cells (erythroblasts) expressing Glycophorin A and CD71. This simple 2D differentiation protocol is performed in two stages. During the first 10 to 12 day stage, STEMdiff™ Hematopoietic Supplement A and then Supplement E1 are added to basal medium to induce cells toward erythroid-biased hematopoietic progenitor cells. At the end of this stage, hematopoietic progenitor cells are easily harvested from the culture supernatant. During the second 14-day stage, cells are further differentiated to erythroid progenitor cells using Supplement E2 and StemSpan™ SFEM II (Day 10 to 24).
The cells typically expand 241-fold +/- 190-fold (95% CI) during the 14-day erythroid differentiation stage and the Day 24 cell population typically contains 74% +/- 5% (95 CI) CD71+GlyA+ erythroblasts. Cells generated using STEMdiff™ Erythroid Kit can be further matured into normoblasts and reticulocytes when moved to appropriate culture conditions for maturation. STEMdiff™ Erythroid Kit has been optimised for differentiation of cells maintained in mTeSR™1, mTeSR™ Plus, and TeSR™-E8™.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').