ContactUsLinkComponent
PancreaCult™ Organoid Growth Medium (Mouse)
PancreaCult™ Organoid Growth Medium (Mouse)
Catalog # STMC06040
CAS Number:  
undefined
PancreaCult™ Organoid Growth Medium (Mouse)
Catalog # STMC06040
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    PancreaCult™ organoid growth medium (mouse)
Specifications
About this item
Cell culture medium for establishment and maintenance of mouse pancreatic exocrine organoids.
PancreaCult™ Organoid Growth Medium (Mouse) is a serum-free, defined cell culture medium for establishment and maintenance of mouse pancreatic exocrine organoids. These organoids, or 'mini-pancreata', provide an in vitro organotypic culture system for studying pancreatic cell biology, disease, and cancer. Organoids grown in PancreaCult™ feature an epithelium expressing genes marking pancreatic stem cells (LGR5), progenitor cells (PDX1, SOX9), and ductal cells (CAR2, MUC1, KRT19).
Pancreatic organoids can be passaged every 3 to 6 days for long-term maintenance and can also be cryopreserved. PancreaCult™ supports mouse pancreatic organoid culture either embedded in Corning® Matrigel® domes or in a dilute Matrigel® suspension. Organoid culture enables convenient in vitro characterisation of the pancreatic epithelium in a physiologically relevant system and reduces the need for animal use.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').