ContactUsLinkComponent
mTeSR™-1 Medium without Phenol Red
mTeSR™-1 Medium without Phenol Red
Catalog # STMC05876
CAS Number:  
undefined
mTeSR™-1 Medium without Phenol Red
Catalog # STMC05876
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    mTeSR™1 without phenol red
Specifications
About this item
Defined, feeder-free maintenance medium for human ES and iPS cells.
mTeSR™1 is the most widely published feeder-free cell culture medium for human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPS cells), with established protocols for applications ranging from derivation to differentiation. It has been used to successfully maintain hundreds of ES and iPS cell lines in over 40 countries, and has supported top pluripotent stem cell publications and researchers. mTeSR™1 is a highly specialised, serum-free, and complete cell culture medium. With pre-screened raw materials that ensure batch-to-batch consistency and robust feeder-free protocols for ES and iPS cell culture, mTeSR™1 provides more consistent cultures with homogeneous, undifferentiated phenotypes. mTeSR™1 is now available without phenol red, for applications in which the presence of phenol red is undesirable.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').