ContactUsLinkComponent
NeuroCult™ Proliferation Supplement (Mouse and Rat)
NeuroCult™ Proliferation Supplement (Mouse and Rat)
Catalog # STMC05701
CAS Number:  
undefined
NeuroCult™ Proliferation Supplement (Mouse and Rat)
Catalog # STMC05701
CAS Number:  
Specifications
  • Pk:
    50 mL
  • Description:
    NeuroCult™ Proliferation supplement (mouse and rat)
Specifications
About this item
Supplement for expansion of mouse and rat neural stem and progenitor cells.
NeuroCult™ Proliferation Supplement (Mouse and Rat) is a standardised, serum-free supplement for the culture of mouse and rat neural stem and progenitor cells. NeuroCult™ Proliferation Supplement (Mouse and Rat) is designed to be combined with NeuroCult™ Basal Medium and appropriate cytokines (not included). NeuroCult™ Proliferation Supplement (Mouse and Rat) is optimised to maintain mouse and rat neural stem cells in culture for extended periods of time without the loss of their self-renewal, proliferation, or differentiation potential. This supplement is also a component of NeuroCult™ Proliferation Kit. NOTE: When preparing Complete NeuroCult™ Proliferation Medium, addition of Human Recombinant EGF is required. When culturing cells obtained from adult mouse or rat, Human Recombinant bFGF and Heparin Solution are also required.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').