ContactUsLinkComponent
MesenCult™ ACF Chondrogenic Differentiation Kit
MesenCult™ ACF Chondrogenic Differentiation Kit
Catalog # STMC05455
CAS Number:  
undefined
MesenCult™ ACF Chondrogenic Differentiation Kit
Catalog # STMC05455
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    MesenCult™ ACF chondrogenic differentiation kit
Specifications
About this item
Animal component-free medium for the differentiation of MSCs into chondrocytes.
MesenCult™-ACF Chondrogenic Differentiation Medium is animal component-free (ACF) and specifically formulated for the in vitro differentiation of primary human mesenchymal stromal cells, and hPSC-derived mesenchymal progenitor cells (MSCs; also known as mesencyhymal stem cells) into chondrogenic lineage cells, including chondrocytes. This medium is suitable for the differentiation of human bone marrow (BM)-, adipose tissue (AT)- and synovium (S)-derived MSCs previously culture-expanded in serum-containing medium (e.g. MesenCult™ Proliferation Kit or animal component-free MesenCult™-ACF Plus Medium. MesenCult™-ACF Chondrogenic Differentiation Medium induces robust chondrogenic differentiation of human MSCs with as few as 3×10⁵ cells and as early as day 14.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').