ContactUsLinkComponent
STEMdiff™ Cardiomyocyte Dissociation Kit
STEMdiff™ Cardiomyocyte Dissociation Kit
Catalog # STMC05025
CAS Number:  
undefined
STEMdiff™ Cardiomyocyte Dissociation Kit
Catalog # STMC05025
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    STEMdiff™ Cardiomyocyte dissociation kit
Specifications
About this item
For dissociation of hPSC-derived cardiomyocytes.
STEMdiff™ Cardiomyocyte Dissociation Kit includes STEMdiff™ Cardiomyocyte Dissociation Medium and STEMdiff™ Cardiomyocyte Support Medium. STEMdiff™ Cardiomyocyte Dissociation Medium can be used to harvest cardiomyocytes that have been differentiated from human pluripotent stem cells (hPSCs) using STEMdiff™ Ventricular Cardiomyocyte Differentiation Kit or STEMdiff™ Atrial Cardiomyocyte Differentiation Kit and maintained in STEMdiff™ Cardiomyocyte Maintenance Medium. STEMdiff™ Cardiomyocyte Support Medium reduces stress on these cardiomyocytes during harvesting and replating, maintaining their viability and functional capacity for downstream applications and analyses.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').