Order Entry
Belgium
Orders LinkContactUsLinkComponent
Formalin solution 10% histological fixative, neutral buffered

Formalin solution 10% histological fixative, neutral buffered

200 L
Catalog # HT501850-200L
Formalin solution 10% histological fixative, neutral buffered
Catalog # HT501850-200L
CAS Number:  50-00-0
Formalin solution 10% histological fixative, neutral buffered

200 L

CAS Number:  50-00-0
Price
962,00 €
Log in for availability
1 * 200 L
Product Details & Documents

Formalin solution, neutral buffered, 10% may be used as a reagent for the primary fixation of tissues for diagnostic applications of an immunoperoxidase method in hematopathology.

Synonyms:  Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Density:1.080 g/cm3
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Room Temperature
Specifications

Specification Test Results

  • Appearance (Colour):
    Colorless
  • Appearance (Turbidity):
    Clear
  • Performance:
    Pass
  • pH:
    6.90 - 7.10

Specifications

  • Pk:
    200 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.