Orders LinkContactUsLinkComponent
STEMdiff™ Midbrain Organoid Differentiation Kit
STEMdiff™ Midbrain Organoid Differentiation Kit
STEMdiff™ midbrain organoid differentiation kit, 1 KIT
Catalog # STMC100-1096
Marketsource
STEMdiff™ Midbrain Organoid Differentiation Kit
Catalog # STMC100-1096
Marketsource
CAS Number:  
STEMdiff™ Midbrain Organoid Differentiation Kit
STEMdiff™ Midbrain Organoid Differentiation Kit
STEMdiff™ midbrain organoid differentiation kit, 1 KIT
CAS Number:  
Price
954,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Cell culture medium kit for robust generation of midbrain neural organoids from human pluripotent stem cells.
Robustly generate three-dimensional, patterned brain organoid cultures from human pluripotent stem cells without matrix embedding by using STEMdiff™ midbrain organoid differentiation kit. When paired with AggreWell™800™, this serum-free cell culture media can prevent organoid fusion and support the generation of over 500 organoids per kit for higher-powered statistical replicates and more detailed longitudinal study. Adapted from protocols, the brain-region-specific organoids generated with this kit are three-dimensional in vitro models with a cellular composition and structural organization representative of the developing human midbrain.
They are consistently sized with reproducible morphology between various cell lines, and provide a reliable model for the study of dopaminergic neurons within a functional 3D system. Midbrain organoids can be combined with striatal organoids generated using the STEMdiff™ Dorsal Forebrain Organoid Differentiation Kit with additional patterning factors to generate midbrain-striatal AssemBloid™ cultures. Organoids can be maintained with STEMdiff™ neural organoid maintenance kit to support long-term culture survival (>50 days) for predictive assays, high-throughput phenotypic screening, and neurotoxicity assays.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    STEMdiff™ midbrain organoid differentiation kit