ContactUsLinkComponent
STEMdiff™ Branching Lung Organoid Maturation Kit
STEMdiff™ Branching Lung Organoid Maturation Kit
Catalog # STMC100-0528
CAS Number:  
undefined
STEMdiff™ Branching Lung Organoid Maturation Kit
Catalog # STMC100-0528
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    STEMdiff™ branching lung organoid maturation kit
Specifications
About this item
Serum-free media for maturation of branching lung organoids from human ES and iPS cells.
Mature and maintain human branching lung organoids in long-term (>28 days) culture using STEMdiff™ branching lung organoid maturation kit. Serum-free STEMdiff™ branching lung organoid maturation kit may be used to reproducibly mature human embryonic stem (ES) and induced pluripotent stem (iPS) cell-derived lung bud organoids generated using STEMdiff™ branching lung organoid kit. Resulting organoids will develop proximal and distal-like branching airway epithelial structures expressing EPCAM, NKX2.1, SOX2, SOX9, MUC1, and P63. When maintained beyond 28 days in a Matrigel® sandwich, there is an increase in expression levels of markers for more mature lung cells, such as SFTPC, SFTPB, and ABCA3.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').