Specifications
- Pk:1 KIT
- Description:STEMdiff™ microglia maturation kit
Specifications
About this item
Maturation kit for the generation of microglia from human ES and iPS cell-derived microglia precursors.
The STEMdiff™ microglia culture system comprises STEMdiff™ microglia differentiation kit and STEMdiff™ microglia maturation kit. Together, these kits are used to differentiate and mature microglia derived from human pluripotent stem cells (hPSCs) using STEMdiff™ hematopoietic kit. Based on the protocol from the laboratory, the resulting cells are a highly pure population of microglia (>80% CD45/CD11b-positive, >50% TREM2-positive microglia; >20% morphologically distinct monocytes or macrophages). Cells derived using these products are versatile tools for modeling neuroinflammation, studying human neurological development and disease, co-culture applications, and toxicity testing.