Hydrocortisone Stock Solutions
Hydrocortisone stock solution (200X) for use with PneumaCult™-ALI, MammoCult™, EpiCult™.
Hydrocortisone stock solution is recommended for use as a cell culture supplement and for the preparation of PneumaCult™, MammoCult™, and EpiCult™ media when culturing airway epithelial cells, mammary primary cells, or tumor cell lines. Hydrocortisone stock solution is provided at a concentration of 96 µg/ml, which is ideal as a 200X stock solution for use with PneumaCult™-ALI, MammoCult™, EpiCult™-B (Human), and EpiCult™-C media. It may also be used for preparing PneumaCult™-ex medium or PneumaCult™-ex plus medium.
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Frequently Bought Together