ContactUsLinkComponent
ReproRNA™ OKSGM
ReproRNA™ OKSGM
Catalog # STMC05931
CAS Number:  
undefined
ReproRNA™ OKSGM
Catalog # STMC05931
CAS Number:  
Specifications
  • Pk:
    12 µG
  • Description:
    ReproRNA™-OKSGM
Specifications
About this item
A non-integrating, self-replicating RNA reprogramming vector for generating iPS cells.
ReproRNA™-OKSGM is a single-stranded RNA replicon vector that contains five reprogramming factors: OCT4, KLF-4, SOX2, GLIS1, and c-MYC, as well as a puromycin-resistance gene. This RNA vector reprograms somatic cells, such as fibroblasts, into induced pluripotent stem (iPS) cells with high efficiency and only requires a single transfection step. When used together with ReproTeSR™ reprogramming medium, the generation of iPS cell colonies can be achieved under feeder-free conditions with superior colony morphology and similar reprogramming efficiency to feeder-based systems. ReproRNA™-derived iPS cell colonies also express markers of undifferentiated cells and retain a normal karyotype. Subsequently, iPS cells generated with ReproRNA™-OKSGM can be maintained in TeSR™ maintenance media (mTeSR™1, TeSR™2, or TeSR™-E8™) and further differentiated into cells of all three germ layers.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').