ContactUsLinkComponent
NeuroCult™ Neuronal Plating Medium
NeuroCult™ Neuronal Plating Medium
Catalog # STMC05713
CAS Number:  
undefined
NeuroCult™ Neuronal Plating Medium
Catalog # STMC05713
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    NeuroCult™ neuronal plating medium
Specifications
About this item
Culture medium for plating dissociated primary tissue-derived neurons for improved survival.
NeuroCult™ neuronal plating medium is a serum-free neuronal basal medium. It is designed for use with BrainPhys™ neuronal medium for the plating and culture of primary tissue-derived neurons.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').