STEMdiff™ Kidney Organoid Kit
Catalog # STMC05160
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:1 KIT
- Description:STEMdiff™ kidney organoid kit
Specifications
About this item
Serum-free medium kit for the culture of kidney organoids from hPSCs.
STEMdiff™ kidney organoid kit is a complete, serum-free cell culture medium system that supports highly efficient and reproducible generation of human pluripotent stem cell (hPSC)-derived kidney organoids in a simple, two-stage differentiation protocol. These kidney organoids are composed of podocytes, proximal tubules, distal tubules, as well as the associated endothelium and mesenchyme. Kidney organoids generated with STEMdiff™ kidney organoid kit are tested for compatibility with phenotypic high-throughput assays such as nephrotoxic compound screening. They also provide a relevant and convenient model for studies related to developmental biology and disease modeling.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').