Orders LinkContactUsLinkComponent
STEMdiff™ Definitive Endoderm Kit
STEMdiff™ definitive endoderm kit, 1 KIT
Catalog # STMC05110
Marketsource
STEMdiff™ Definitive Endoderm Kit
Catalog # STMC05110
Marketsource
CAS Number:  
STEMdiff™ Definitive Endoderm Kit
STEMdiff™ Definitive Endoderm Kit
STEMdiff™ definitive endoderm kit, 1 KIT
CAS Number:  
Price
520,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Defined animal component-free medium for the differentiation of human ES and iPS cells to definitive endoderm.
STEMdiff™ definitive endoderm kit is a complete, serum- and animal component-free medium and supplement kit that supports highly efficient differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells to definitive endoderm cells. Cells differentiated using STEMdiff™ definitive endoderm kit express high levels of endoderm markers, including CD184 (CXCR4), SOX17, FOXA2, and c-KIT, and lack expression of ectoderm, mesoderm, and pluripotency markers. The definitive endoderm cells produced using this kit are multipotent and capable of further differentiation towards cells of the pancreatic, intestinal, pulmonary, and hepatic lineages, thus providing a robust tool for developmental studies, disease modeling, and drug discovery.​ This kit is optimised for differentiation of cells maintained in mTeSR™1. For differentiation of cells maintained in TeSR™-E8™, please see the STEMDiff™ Definitive Endoderm Kit (TeSR™-E8™ optimised).
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    STEMdiff™ definitive endoderm kit
Specifications