MegaCult™-C Collagen and Medium with Lipids
Catalog # STMC04974
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:1 KIT
- Description:MegaCult™-C collagen and medium with lipids
Specifications
About this item
Kit containing collagen and medium with lipids, without cytokines, for human and mouse CFU-Mk assays.
MegaCult™-C collagen and medium with lipids kit includes medium and collagen solution for colony-forming unit (CFU) assays of human or mouse megakaryocyte progenitor cells (CFU-Mk). The addition of cytokines is required.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').