Orders LinkContactUsLinkComponent
Human LDL
Human LDL
Human LDL, 5 mg
Catalog # STMC02698
Marketsource
Human LDL
Catalog # STMC02698
Marketsource
CAS Number:  
Human LDL
Human LDL
Human LDL, 5 mg
CAS Number:  
Price
692,00 €
Log in for availability
1 * 5 mg
Product Details & Documents
Low-density lipoproteins pre-screened for hematopoietic cell growth.
This product is a human plasma-derived low-density lipoprotein (LDL)-based culture supplement that has been pre-screened for the culture, expansion, and colony assay of human hematopoietic and non-hematopoietic cells in serum-free culture media. It promotes the proliferation and survival of human hematopoietic and other progenitor cells in culture, resulting in increased cell output in expansion cultures and increased colony numbers and/or size in colony assays.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Pk:
    5 mg
  • Description:
    Human LDL
Specifications