Order Entry
Belgium
Orders LinkContactUsLinkComponent
Phenol

Phenol

Supplier:   CPACHEM
Phenol
Supplier:  CPACHEM
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
  • Density:1 g/cm³
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 101,00 €
    101,00 €
    1 * 100 mL
       101,00 €

  • 500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 356,00 €
    356,00 €
    1 * 500 mL
       356,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.