Sodium chloride
Catalog # ACSDZ15081092.1L
Supplier: CPACHEM
New Product
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:58.44 g/mol
Boiling Pt:100 °C
Density:1 g/cm³
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477