Order Entry
Belgium
ContactUsLinkComponent
Formalin solution 10%, neutral buffered, Sigma-Aldrich®
Formalin solution 10%, neutral buffered, Sigma-Aldrich®
Catalog # HT501640-19L
CAS Number:  
Formalin solution 10%, neutral buffered, Sigma-Aldrich®
Catalog # HT501640-19L
CAS Number:  

Specifications

  • Pk:
    19 L
  • Seq:
    5282

Specifications

Specification Test Results

  • Appearance (Colour):
    Colorless
  • Appearance (Turbidity):
    Clear
  • Performance:
    Pass
  • pH:
    6.90 - 7.10
  • Expiration Date Period:
    30 months

Specification Test Results

About this item

 :
Density:1.080 g/cm3
 :M-Clarity™ quality level = MQ200
 :30.03 g/mol
Storage Temperature:Room Temperature
CAS Number:50-00-0
Formula:CH₂O
MDL Number:MFCD00003274