Order Entry
Belgium
ContactUsLinkComponent
DMEM Low Glucose with Stable Glutamine with Sodium Pyruvate
DMEM Low Glucose with Stable Glutamine with Sodium Pyruvate
Catalog # A0101-500
Supplier:  Biowest
CAS Number:  
DMEM Low Glucose with Stable Glutamine with Sodium Pyruvate
Catalog # A0101-500
Supplier:  Biowest
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Pk:
    500 mL
  • Description:
    DMEM, low glucose, L-glutamine, pyruvate
  • Sterility:
    Sterile filtered
  • Shipping conditions:
    Room temperature or controlled temperature
  • Storage temperature:
    2...8 °C
  • Shelf life:
    24 months

Specifications

About this item

Dulbecco's Modified Eagle medium (DMEM) is formulated to preserve and maintain the growth of a wide range of mammalian cells. Supplemented with fetal calf serum, DMEM also allows the proliferation of mouse embryonic stem cells.

  • This reference is formulated with stable glutamine