Order Entry
Belgium
ContactUsLinkComponent
Haematoxylin solution Gill II, Sigma-Aldrich®
Corrosive
Haematoxylin solution Gill II, Sigma-Aldrich®
Catalog # GHS2128-4L
CAS Number:  
Corrosive
Haematoxylin solution Gill II, Sigma-Aldrich®
Catalog # GHS2128-4L
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Appearance (Turbidity):
    Clear
  • Appearance(Colour)
    Reddish-Purple:
    Conforms
  • Appearance (Form):
    Solution
  • Performance:
    Conforms
  • Expiration Date Period:
    24 months

Specification Test Results

About this item

 :
 :302.28 g/mol
 :M-Clarity™ quality level = MQ500 (more info.)
 :GHS05
Storage Temperature:Room Temperature
CAS Number:517-28-2
 :208-237-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00078111