ContactUsLinkComponent
AESCULAP® Acculan 4 Dermatome
AESCULAP® Acculan 4 Dermatome
Catalog # AESCGA340
Supplier:  Aesculap
AESCULAP® Acculan 4 Dermatome
Catalog # AESCGA340
Supplier:  Aesculap
Specifications
  • Description:
    Acculan 4 dermatome
  • Pk:
    1
Specifications
About this item
The acculan 4 dermatome is powerful and simple to use. It combines reliability, longevity and ergonomics. A machine that will inspire you.
  • Keyless adjustment of cutting thickness and width
  • Cutting thickness: 0,2 - 1,2 mm
  • Cutting width: max. 78 mm
  • Golden marked operating elements
The titanium housing and the ECCOS storage solution acculan 4 is suitable for mechanical cleaning, even with alkaline cleaning media.
Regular servicing and preventive maintenance is essential to ensure the reliability of motor systems over a long service life. The service label on each acculan 4 machine clearly shows when the next preventive maintenance is due.